Learn More
Eppendorf Copper Water Tray and 4 Copper Shelves
Shop All Eppendorf Products
Click to view available options
Material:
Copper
Unit Size:
Each
Description
- Specifically designed for 170 L incubators, this set facilitates a favorable environment for cell cultures by utilizing the excellent thermal conductivity of copper.
- The water tray helps maintain optimal humidity levels within the incubator, crucial for cell growth and viability.
- The four copper shelves provide ample space to organize samples efficiently and allow for uniform distribution of heat throughout the chamber. Copper's superior heat transfer properties ensure stable and consistent temperature conditions inside the incubator, promoting accurate and reliable results in your experiments.
- Upgrade your incubator with this set to benefit from enhanced heat distribution, improved temperature stability, and better humidity control.
Specifications
Specifications
| For Use With (Equipment) | 170 L Incubator |
| Material | Copper |
| Type | Water Tray and Shelves Set |