missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) tetrafluoroborate hexahydrate, 98%

Product Code. 11372658
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372658 500 g 500g
11382658 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372658 Supplier Thermo Scientific Alfa Aesar Supplier No. 026127.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72259-10-0 | B2CuF8H12O6 | 345.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72259-10-0
Molecular Formula B2CuF8H12O6
Molecular Weight (g/mol) 345.24
MDL Number MFCD05863735
InChI Key FBYIXFBIARQMJJ-UHFFFAOYSA-N
Synonym copper ii tetrafluoroborate hexahydrate, copper 2+ hexahydrate ditetrafluoroborate, copper ditetrafluoroborate hexahydrate, copper 2+ tetrafluoroborate-water 1/2/6, copper 2+ ion hexahydrate ditetrafluoroborate, borate 1-,tetrafluoro-, copper 2+ 2:1 , hexahydrate 9ci
PubChem CID 15774553
IUPAC Name copper(2+) bis(tetrafluoroboranuide) hexahydrate
SMILES O.O.O.O.O.O.[Cu++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Color Blue
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 998%
Solubility Information Soluble in water.
Formula Weight 345.25 (237.15 Anhydrous)
Sensitivity Hygroscopic
Appearance Blue crystals
Chemical Name or Material Copper(II) tetrafluoroborate hexahydrate, hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.