accessibility menu, dialog, popup

UserName

BFFFFBFFFFH2OH2OH2OH2OH2OH2OCu

CAS RN 72259-10-0

IUPAC Name: copper(2+) bis(tetrafluoroboranuide) hexahydrate
Synonyms: copper(2+) hexahydrate ditetrafluoroborate
Molecular Weight (g/mol): 345.24
Molecular Formula: B2CuF8H12O6
InChi Key: FBYIXFBIARQMJJ-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Cu++].F[B-](F)(F)F.F[B-](F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results