missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) 2,4-pentanedionate, 98%

CAS: 13395-16-9 | C10H14CuO4 | 261.76 g/mol

£22.80 - £240.00

Chemical Identifiers

CAS 13395-16-9
Molecular Formula C10H14CuO4
Molecular Weight (g/mol) 261.76
MDL Number MFCD00000016
InChI Key QYJPSWYYEKYVEJ-FDGPNNRMSA-L
Synonym Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
IUPAC Name copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11462737
SDS View Documents
Thermo Scientific Alfa Aesar
A14920.18
50 g
£22.80
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11314549
SDS View Documents
Thermo Scientific Alfa Aesar
A14920.22
100 g
ONLINE EXCLUSIVE PRICE
£37.50
£33.65 / 100g
Save £3.85
10% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11452737
SDS View Documents
Thermo Scientific Alfa Aesar
A14920.0B
1000 g
£240.00
1000g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Copper(II) 2,4-pentanedionate is used as a catalyst for carbene transfer reactions, coupling reactions and Michael addition reactions. It is also used as a catalyst for the polymerization of olefins and transesterification reactions as well as a PVC stabilizer. Further, it is used as a curing agent for epoxy resins, acrylic adhesives, silicone rubbers. In addition to this, it is used as a lubricant additive and paint drier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) 2,4-pentanedionate is used as a catalyst for carbene transfer reactions, coupling reactions and Michael addition reactions. It is also used as a catalyst for the polymerization of olefins and transesterification reactions as well as a PVC stabilizer. Further, it is used as a curing agent for epoxy resins, acrylic adhesives, silicone rubbers. In addition to this, it is used as a lubricant additive and paint drier.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

13395-16-9
261.76
QYJPSWYYEKYVEJ-FDGPNNRMSA-L
copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
C10H14CuO4
MFCD00000016
Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

13395-16-9
160°C (10 mmHg)
C10H14CuO4
4157957
Insoluble in water.
[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
261.76
98%
∼286°C (decomposition)
Weak
MFCD00000016
Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
QYJPSWYYEKYVEJ-FDGPNNRMSA-L
copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
261.76
Copper(II) 2,4-pentanedionate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335-H360-H500

missing translation for 'einecsNumber' : 236-477-9

missing translation for 'rtecsNumber' : GL6520000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.