missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) 2,4-pentanedionate, 98%

Product Code. 11452737
Change view
Click to view available options
Quantity:
50 g
100 g
1000 g
Unit Size:
1000g
100g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452737 1000 g 1000g
11462737 50 g 50g
11314549 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452737 Supplier Thermo Scientific Alfa Aesar Supplier No. A14920.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13395-16-9 | C10H14CuO4 | 261.76 g/mol

Copper(II) 2,4-pentanedionate is used as a catalyst for carbene transfer reactions, coupling reactions and Michael addition reactions. It is also used as a catalyst for the polymerization of olefins and transesterification reactions as well as a PVC stabilizer. Further, it is used as a curing agent for epoxy resins, acrylic adhesives, silicone rubbers. In addition to this, it is used as a lubricant additive and paint drier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) 2,4-pentanedionate is used as a catalyst for carbene transfer reactions, coupling reactions and Michael addition reactions. It is also used as a catalyst for the polymerization of olefins and transesterification reactions as well as a PVC stabilizer. Further, it is used as a curing agent for epoxy resins, acrylic adhesives, silicone rubbers. In addition to this, it is used as a lubricant additive and paint drier.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13395-16-9
Molecular Formula C10H14CuO4
Molecular Weight (g/mol) 261.76
MDL Number MFCD00000016
InChI Key QYJPSWYYEKYVEJ-FDGPNNRMSA-L
Synonym Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
IUPAC Name copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point ∼286°C (decomposition)
Boiling Point 160°C (10 mmHg)
Odor Weak
Quantity 1000 g
Beilstein 4157957
Solubility Information Insoluble in water.
Formula Weight 261.76
Percent Purity 98%
Chemical Name or Material Copper(II) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.