missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) bromide-dimethyl sulfide complex, 99%

Product Code. 10053813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10053813 25 g Glass bottle 25g
10490291 10 g Glass bottle 10g
10257023 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10053813 Supplier Thermo Scientific Acros Supplier No. 213180250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54678-23-8 | C2H6BrCuS | 205.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54678-23-8
Molecular Formula C2H6BrCuS
Molecular Weight (g/mol) 205.58
MDL Number MFCD00043295
InChI Key PMHQVHHXPFUNSP-UHFFFAOYSA-M
Synonym copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i
PubChem CID 2724234
SMILES [Cu]Br.CSC

Specifications

Melting Point 132°C
Color Beige-Gray to White or Green
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodometry)
Packaging Glass bottle
Linear Formula (CH3)2S·CuBr
Quantity 25 g
IUPAC Name (methylsulfanyl)methane; bromocopper
Formula Weight 205.58
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Copper(I) bromide-dimethyl sulfide complex, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.