missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) acetate, 95%

Product Code. 10659404 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10659404

Brand: Thermo Scientific Acros 378810010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 598-54-9 | C2H3CuO2 | 122.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-54-9
Molecular Formula C2H3CuO2
Molecular Weight (g/mol) 122.59
MDL Number MFCD00058908
InChI Key RFKZUAOAYVHBOY-UHFFFAOYSA-M
Synonym copper i acetate, cuprous acetate, copper 1+ acetate, acetic acid, copper 1+ salt, unii-9u3n93lmzb, copper acetate, 9u3n93lmzb, acetic acid, copper 1+ salt 1:1, copper 1 acetate
PubChem CID 69023
IUPAC Name copper(1+);acetate
SMILES CC(=O)[O-].[Cu+]

Specifications

Melting Point 250.0°C
Color White to Yellow
Physical Form Powder
Quantity 1 g
Assay Percent Range 48.7% min. (Cu)
Linear Formula CH3C(O)OCu
Merck Index 15, 2648
Solubility Information Solubility in water: reacts
Formula Weight 122.59
Percent Purity 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Copper(I) acetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.