CAS RN 598-54-9
IUPAC Name:
λ¹-copper(1+) acetate
Synonyms:
λ¹-copper(1+) acetate
Molecular Weight (g/mol):
122.59
Molecular Formula:
C2H3CuO2
InChi Key:
RFKZUAOAYVHBOY-UHFFFAOYSA-M
SMILES:
[Cu+].CC([O-])=O
1
–
1
of
1
results
1
Copper(I) acetate, 95%
CAS: 598-54-9 Molecular Formula: C2H3CuO2 Molecular Weight (g/mol): 122.59 MDL Number: MFCD00058908 InChI Key: RFKZUAOAYVHBOY-UHFFFAOYSA-M Synonym: copper i acetate,cuprous acetate,copper 1+ acetate,acetic acid, copper 1+ salt,unii-9u3n93lmzb,copper acetate,9u3n93lmzb,acetic acid, copper 1+ salt 1:1,copper 1 acetate PubChem CID: 69023 IUPAC Name: copper(1+);acetate SMILES: CC(=O)[O-].[Cu+]