missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) acetate, 95%

Product Code. 10198412 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10198412 5 g Glass bottle 5g
10659404 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10198412 Supplier Thermo Scientific Acros Supplier No. 378810050
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 598-54-9 | C2H3CuO2 | 122.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-54-9
Molecular Formula C2H3CuO2
Molecular Weight (g/mol) 122.59
MDL Number MFCD00058908
InChI Key RFKZUAOAYVHBOY-UHFFFAOYSA-M
Synonym copper i acetate, cuprous acetate, copper 1+ acetate, acetic acid, copper 1+ salt, unii-9u3n93lmzb, copper acetate, 9u3n93lmzb, acetic acid, copper 1+ salt 1:1, copper 1 acetate
PubChem CID 69023
IUPAC Name copper(1+);acetate
SMILES CC(=O)[O-].[Cu+]

Specifications

Melting Point 250.0°C
Color White to Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 48.7% min. (Cu)
Linear Formula CH3C(O)OCu
Merck Index 15, 2648
Solubility Information Solubility in water: reacts
Formula Weight 122.59
Percent Purity 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Copper(I) acetate
Show More Show Less
compliance-icons
Product Identifier
  • Copper(I) acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.