missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
COPE Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Tekniske data
| Antigen | COPE |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
18696651
|
Novus Biologicals
NBP2-91992-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609930
|
Novus Biologicals
NBP2-91992-0.1ml |
0.1 mL |
£359.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
COPE Polyclonal antibody specifically detects COPE in Human, Mouse, Rat samples. It is validated for Western BlotTekniske data
| COPE | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 11316 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| coatomer epsilon subunit, coatomer protein complex, subunit epsilon, coatomer subunit epsilon, epsilon coat protein, Epsilon-coat protein, epsilon-COP, FLJ13241 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-308 of human COPE (NP_009194.2). MAPPAPGPASGGSGEVDELFDVKNAFYIGSYQQCINEAQRVKLSSPERDVERDVFLYRAYLAQRKFGVVLDEIKPSSAPELQAVRMFADYLAHESRRDSIVAELDREMSRSVDVTNTTFLLMAASIYLHDQNPDAALRALHQGDSLECTAMTVQILLKLDRLDLARKELKRMQDLDEDATLTQLATAWVSLATGGEKLQDAYYIFQEMADKCSPTLLLLNGQAACHMAQGRWEAAEGLLQEALDKDSGYPETLVNLIVLSQHLGKPPEVTNRYLSQLKDAHRSHPFIKEYQAKENDFDRLVLQYAPSA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel