missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPE Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-91992-0.1ml
This item is not returnable.
View return policy
Description
COPE Polyclonal antibody specifically detects COPE in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| COPE | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| coatomer epsilon subunit, coatomer protein complex, subunit epsilon, coatomer subunit epsilon, epsilon coat protein, Epsilon-coat protein, epsilon-COP, FLJ13241 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-308 of human COPE (NP_009194.2). MAPPAPGPASGGSGEVDELFDVKNAFYIGSYQQCINEAQRVKLSSPERDVERDVFLYRAYLAQRKFGVVLDEIKPSSAPELQAVRMFADYLAHESRRDSIVAELDREMSRSVDVTNTTFLLMAASIYLHDQNPDAALRALHQGDSLECTAMTVQILLKLDRLDLARKELKRMQDLDEDATLTQLATAWVSLATGGEKLQDAYYIFQEMADKCSPTLLLLNGQAACHMAQGRWEAAEGLLQEALDKDSGYPETLVNLIVLSQHLGKPPEVTNRYLSQLKDAHRSHPFIKEYQAKENDFDRLVLQYAPSA | |
| 0.1 mL | |
| Signal Transduction | |
| 11316 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction