missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cooked Meat Medium (Bottled)

Product Code. 12383793
Change view
Click to view available options
Quantity:
100/Pk.
24/Pk.
Unit Size:
Each
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12383793 24/Pk. Each
12743586 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12383793 Supplier Thermo Scientific™ Supplier No. BO0212E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Grow and maintain aerobic and anaerobic organisms with these nutritious, ready prepared bottles.

Grow and maintain aerobic and anaerobic organisms with Thermo Scientific ™ Oxoid™ Cooked Meat Medium ready prepared bottles. The medium contains heart tissue, beef extracts, peptone and glucose for the cultivation of anaerobic and aerobic organisms. It has the ability to initiate bacterial growth from very small inocula and to maintain the viability of cultures over long periods of time1. Mixed cultures of bacteria survive in Cooked Meat Medium without displacing the slower growing organisms.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared bottles of Cooked Meat Medium.

  • Suitable for primary growth and maintenance of aerobic and anaerobic organisms.
  • Preserves the viability of organisms over a long period of time and is useful in maintaining anaerobic stock organisms.
  • Convenient: the pre-filled bottles are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months

The addition of glucose to the formulation allows rapid, heavy growth of anaerobic bacteria in a short time and leads to a more rapid identification of important anaerobes. The improved growth also enhances GLC identification of anaerobic bacteria.

The improved clarity of the supernatant broth permits earlier detection of growth especially when combined with the increased growth of most organisms. Slower growing isolates will yield detectable growth within 45 hours of incubation.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw, slight haze
Content And Storage 2 to 25°C
Description Cooked Meat Medium (Bottled)
Format Universal Bottles
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 10.0g + 0.3g
Packaging Type Twenty four universal bottles with screw cap closures in a box
Quantity 24/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.