missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Controlled Rate Freezer

Product Code. 17215546 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
1 x SBS Microplate
16 x 0.5 mL CBS High Security Straws
18 x 0.3 mL IMV Straws
55 x 1.8 mL Cryovials (0.5 mL Maximum Fill)
55 x 1.8 mL Cryovials (1 mL Maximum Fill)
Flat Plate for Items or Vessels
Plate for 10 x 0.5 mL CBS High Security Straws, 12 x 1.8 mL Cryovials (0.5 mL Maximum Fill)
Plate for 12 x 0.3 mL IMV Straws, 12 x 1.8 mL Cryovials (0.5 mL Maximum Fill)
PC Programmability:
CRF-1 User Software
User Software
Plug Type:
EU, UK
EU, UK, US
Temperature Range:
-100°C
-80°C
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds PC Programmability Plug Type Temperature Range unitSize
17215546 55 x 1.8 mL Cryovials (0.5 mL Maximum Fill) User Software EU, UK -80°C Each
17295536 16 x 0.5 mL CBS High Security Straws User Software EU, UK -100°C Each
17205546 18 x 0.3 mL IMV Straws User Software EU, UK, US -100°C Each
17245546 55 x 1.8 mL Cryovials (1 mL Maximum Fill) User Software EU, UK, US -80°C Each
17235546 1 x SBS Microplate CRF-1 User Software EU, UK, US -80°C Each
17225546 Flat Plate for Items or Vessels User Software EU, UK, US -100°C Each
17255546 Plate for 10 x 0.5 mL CBS High Security Straws, 12 x 1.8 mL Cryovials (0.5 mL Maximum Fill) User Software EU, UK -100°C Each
17265546 Plate for 12 x 0.3 mL IMV Straws, 12 x 1.8 mL Cryovials (0.5 mL Maximum Fill) User Software EU, UK, US -100°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 17215546

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Liquid nitrogen free and cryogen free controlled rate freezer for research into the cryopreservation of a wide range of material including: embryos, stem cells, mammalian cells, spermatozoa, antibodies, tissue sections and rodent organs.

The Fisherbrand™ Controlled Rate Freezer is a liquid nitrogen free and cryogen free controlled rate freezer that brings accuracy, precision and reproducibility to biological cryopreservation.

Unlike conventional liquid nitrogen based controlled rate cooling equipment, the Fisherbrand™ Controlled Rate Freezer poses no contamination risk and can be used in cleanrooms and barrier facilities. It fits neatly and quietly on a bench-top and its performance in terms of cell viability after freezing is comparable or better than standard liquid nitrogen freezers. As alcohol is not used, there is also no potential fire risk. The Fisherbrand™ Controlled Rate Freezer will cool down to -100°C with straws.

The cooling rate is precisely controlled, ensuring accuracy and reproducibility throughout the freezing profile, especially for the important nucleation/seeding phase. This ensures optimal recovery of cells upon thawing. Operation is simple and can be carried out with or without a PC; data can be logged via PC software and cooling profiles are directly displayed. Different cooling profiles are available from a drop down menu and customised profiles can be written.

Main applications: Fisherbrand™ Controlled Rate Freezer is highly versatile and can be used for the cryopreservation research of a wide range of samples in cryovials, straws, bags, microplates and Matrix-96-well block plates in the following areas:

  • Transgenic embryos research
  • Stem cell research
  • Clinical and research samples, e.g. lymphocytes and tissue cell lines in conventional cryovials
  • Various mammalian cells including cardiomyocytes, adipose, liver and muscle
  • Cord blood derived stem cells
  • Adherent cells and stem cells in microplates
  • Cell suspensions in numbered/barcoded arrays
  • Robotic integration – the Fisherbrand™ Controlled Rate Freezer has also successfully been integrated into robotic systems
  • Suitable for applications in veterinarian IVF

Key benefits and features:

  • Accurate and reproducible control of cooling rates and sample temperatures
  • Easy to use and samples can be nucleated/seeded in-situ
  • Linear and non-linear cooling profiles
  • Low running costs: estimated at 1% of liquid nitrogen controlled rate freezing
  • Temperature remains at -100°C at the end of cycle for straw applications until freezer is switched off
  • Uninterruptible Power Supply (UPS): complete cycle run if power fails (supplied as an optional accessory)
  • CE marked (laboratory use)
  • Servicing and calibration available
  • 3 year warranty

Specifications:

  • Overall Size: W 36.9 cm, D 26.6 cm, H 45.3 cm (with lid)
  • Operating temperature (environment): 20°C to 30°C
  • A number of fixed plates available (standard straws, high security straws, cryovials, cryocyte bags and microplates)
  • Freezer can be run as a stand alone pre-programmed unit or connected to a control PC (computer with serial port and control software and Windows)
  • Software allowing configuration of existing profiles, creation of new customer profiles, data logging and reporting. All data is logged in an encrypted and Excel readable form.
  • Standard, pre-set cooling profiles available
  • Backup electrical supply: 750 VA UPS capable of running freezer for 3 hour cycle in the case of an electrical power failure
  • Cleaning: Seamless, no dirt traps, may be cleaned using standard wipes
TRUSTED_SUSTAINABILITY

Specifications

Type Cryogenic Freezer
Temperature Range -80°C
Chamber Material Aluminum
Certifications/Compliance RoHS, CE, UKCA
Style Benchtop
Color Gray
Warranty 3 Years
Defrost Automatic
Dimensions (DxWxH) Exterior 264 x 367 x 445 mm
PC Programmability User Software
Amperage 2.5 A
Plug Type EU, UK
Electrical Requirements Mains Supply: 100/240 V, 50/60 Hz, Unit Supply: 24 V DC
Wattage 150 W
Energy Usage 150 W
Compressor CFC-free
Weight (Metric) 9.8 kg
Holds 55 x 1.8 mL Cryovials (0.5 mL Maximum Fill)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.