missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 32 Mouse, Clone: GJB1/1753, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £423.29
Specifications
| Antigen | Connexin 32 |
|---|---|
| Clone | GJB1/1753 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
Specifications
| Connexin 32 | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 2705 | |
| Recombinant human GJB1 protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 27-32kDa |
| GJB1/1753 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| XP_016884897 | |
| GJB1 | |
| IgG | |
| Protein A/G Purified | |
| Cytoplasmic and Cell Surface | |
| MCF-7 Cells. Liver, Kidney, Stomach or Tonsil |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title