missing translation for 'onlineSavingsMsg'
Learn More

Connexin 32 Mouse, Clone: GJB1/1753, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 2705-MSM3-P1

 View more versions of this product

Product Code. 15930098

  • £423.29 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Connexin 32
Cytoplasmic and Cell Surface
GJB1/1753
Unconjugated
XP_016884897
Mouse
27-32kDa
Protein A/G Purified
RUO
2705
IgG
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
GJB1
Recombinant human GJB1 protein
MCF-7 Cells. Liver, Kidney, Stomach or Tonsil
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.