missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 32/GJB1 Antibody (SR1532), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £358.00
Specifications
| Antigen | Connexin 32/GJB1 |
|---|---|
| Clone | SR1532 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625384
|
Novus Biologicals
NBP3-22393-100ul |
100 μg |
£358.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608554
|
Novus Biologicals
NBP3-22393-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Connexin 32/GJB1 Monoclonal antibody specifically detects Connexin 32/GJB1 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
| Connexin 32/GJB1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2705 | |
| IgG | |
| Affinity purified |
| SR1532 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CMTX, Connexin-32, Cx32, CX32beta 1, 32kD (connexin 32, Charcot-Marie-Toothneuropathy, X-linked), gap junction beta-1 protein, gap junction protein, beta 1, 32kDa, gap junction protein, beta 1, 32kDa (connexin 32), gap junction protein, beta 1, 32kDa (connexin 32, Charcot-Marie-Toothneuropathy, X-linked) | |
| A synthesized peptide derived from human Connexin 32/GJB1 (Uniprot #: P08034) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title