missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 32/GJB1 Antibody (SR1532), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22393-50ul
This item is not returnable.
View return policy
Description
Connexin 32/GJB1 Monoclonal antibody specifically detects Connexin 32/GJB1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| Connexin 32/GJB1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2705 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| SR1532 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
| CMTX, Connexin-32, Cx32, CX32beta 1, 32kD (connexin 32, Charcot-Marie-Toothneuropathy, X-linked), gap junction beta-1 protein, gap junction protein, beta 1, 32kDa, gap junction protein, beta 1, 32kDa (connexin 32), gap junction protein, beta 1, 32kDa (connexin 32, Charcot-Marie-Toothneuropathy, X-linked) | |
| A synthesized peptide derived from human Connexin 32/GJB1 (Uniprot #: P08034) | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction