Learn More
Binder Connection Kit VD VP1-VP2
Product Code. 30503398 Shop All BINDER Products
Change view
Click to view available options
For Use With:
Vacuum Pump VP1 and VP2
Unit Size:
Each
Product Code. For Use With unitSize
30503398 Vacuum Pump VP1 and VP2 Each
1 options
Product Code. 30503398 Supplier Binder Supplier No. 80122613
For vacuum pump VP1 and VP2.
  • Consisting of plastic hose 1.9 m long, clamping ring, universal centering ring and small flange adapter 1/2 in. in diameter (DN 16) to hose nozzle 3/8 in. in diameter (DN 10).
Specifications
For Use With Vacuum Pump VP1 and VP2
IP Rating IP40
Type Pump Connection Kit