missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Separating Funnel, conical shape, with PTFE key

Product Code. 10243512 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10243512 1000 mL Each
10580773 100 mL Each
10426332 500 mL Each
10658013 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10243512 Supplier DWK Life Sciences Supplier No. 1064809
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The separating funnel enables the separation of immiscible liquids (liquid-liquid extraction).

A separating funnel has the shape of a pointed flask with a drain cock at its tip. The separating funnel is used for the liquid-liquid extraction of two immiscible liquids (substance separation process) and can thus also be used for cleaning processes. The liquid with the higher density collects in the lower part of the vessel and can be drained via the drain cock. The printed scale allows an exact volume determination.

  • Conical shape
  • With PTFE key
  • Fired-on, highly durable white print
  • Easy-to-read scale with graduation marks to estimate the content
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Diameter (Metric) Stem 13 mm
Material Boro 3.3, PTFE, Polypropylene
Autoclavable Autoclavable
For Use With (Application) Phase separation; Liquid-liquid extraction; Measuring liquid amounts
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.