Learn More
Agilent BioTek Complete Dispense and Waste Systems
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
50 TS Microplate Washer, Agilent BioTek Microplate Washer and Reader Instruments
50 TS with Automated Buffer Switching Capabilities, Agilent BioTek Microplate Washer and Reader Instruments
Unit Size:
Each
Description
4070077 includes: 3 Supply Bottles with Filter, 4 L; 1 Waste Bottle, 4 L; 2 Racks, and Tubing, 4070078 includes: 2 Dispense Bottles with Filter, 4L; 1 Waste Bottle, 4L ; 2 Racks, and Tubing
Specifications
Specifications
| Type | Waste System |
| For Use With (Equipment) | 50 TS with Automated Buffer Switching Capabilities, Agilent BioTek Microplate Washer and Reader Instruments |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido