Learn More
Agilent BioTek Complete Direct Drain Waste Systems
GREENER_CHOICE
Product Code. 15993300 Shop All Agilent BioTek Products
Click to view available options
Unit Size:
Each
Type:
Complete Direct Drain Waste System
Product Code. 15993300
Brand: Agilent BioTek 1170562S
Drains into sink instead of bottles
  • Compatible with Agilent BioTek 405 and 406 Series Microplate Washer Systems
  • Eliminates the need of attending to and emptying waste bottles
  • Saves space on the workbench
TRUSTED_SUSTAINABILITY
Specifications
Includes Direct drain module (115V/230V), standard or high-flow vacuum pump, tubing, holder for two 4L bottles
For Use With (Equipment) Agilent BioTek 406 Series Microplate Washer System