missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor H-related 5/CFHR5 Antibody (3A10A5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52441-0.025mg
This item is not returnable.
View return policy
Description
Complement Factor H-related 5/CFHR5 Monoclonal specifically detects Complement Factor H-related 5/CFHR5 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| Complement Factor H-related 5/CFHR5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| CFHL5factor H-related protein 5, complement factor H-related 5, FHR-5complement factor H-related protein 5, FHR5MGC133240, FLJ10549 | |
| Mouse | |
| 64.4 kDa | |
| 0.025 mg | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 81494 | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3A10A5 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CFHR5 | |
| Purified recombinant fragment of human CFHR5 (AA: 344-569) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction