missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor H-related 5/CFHR5 Antibody (3A10A5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£184.00 - £463.00
Specifications
| Antigen | Complement Factor H-related 5/CFHR5 |
|---|---|
| Clone | 3A10A5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18257160
|
Novus Biologicals
NBP2-52441 |
0.1 mg |
£463.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668528
|
Novus Biologicals
NBP2-52441-0.025mg |
0.025 mg |
£184.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Factor H-related 5/CFHR5 Monoclonal specifically detects Complement Factor H-related 5/CFHR5 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Complement Factor H-related 5/CFHR5 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| CFHL5factor H-related protein 5, complement factor H-related 5, FHR-5complement factor H-related protein 5, FHR5MGC133240, FLJ10549 | |
| CFHR5 | |
| IgG2a | |
| Protein A or G purified |
| 3A10A5 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 81494 | |
| Purified recombinant fragment of human CFHR5 (AA: 344-569) expressed in E. Coli. | |
| Primary | |
| 64.4 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title