missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C9 Antibody (WU13-15), Novus Biologicals™
Human Monoclonal Antibody
£177.00 - £581.00
Specifications
| Antigen | ComplementComponentC9 |
|---|---|
| Clone | WU13-15 |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18270832
|
Novus Biologicals
NBP2-62803 |
100 μg |
£581.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18230504
|
Novus Biologicals
NBP2-62803-0.025MG |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Component C9 Monoclonal specifically detects Complement Component C9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.Specifications
| ComplementComponentC9 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay | |
| Unconjugated | |
| Human | |
| PBS containing 0.1% bovine serum albumin with 0.02% Sodium Azide | |
| 735 | |
| Neoepitope on the 61 kDa complement component C9, integrated in the terminal complement complex (TCC). | |
| Primary |
| WU13-15 | |
| Monoclonal | |
| Purified | |
| Human | |
| complement component 9, complement component C9 | |
| C9 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title