missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C9 Antibody (WU13-15), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP2-62803
This item is not returnable.
View return policy
Description
Complement Component C9 Monoclonal specifically detects Complement Component C9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.
Specifications
| ComplementComponentC9 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay | |
| complement component 9, complement component C9 | |
| Human | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), Western Blot | |
| WU13-15 | |
| Unconjugated | |
| PBS containing 0.1% bovine serum albumin with 0.02% Sodium Azide | |
| C9 | |
| Neoepitope on the 61 kDa complement component C9, integrated in the terminal complement complex (TCC). | |
| 100 μg | |
| 735 | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction