missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C2b Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £377.00
Specifications
| Antigen | Complement Component C2b |
|---|---|
| Clone | 269716 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18759547
![]() |
R&D Systems
MAB1936-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18130161
![]() |
R&D Systems
MAB1936 |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Component C2b Monoclonal specifically detects Complement Component C2b in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Complement Component C2b | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P06681 | |
| 717 | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Component C2 Ala21-Leu752 Accession # P06681 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Complement Component C2b in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) C1r, rhC1s, rhC3r, rhC5a, rhFactor B, or rhFactor I is observed. In Western blots, this antibody recognizes rhC2 and rhC2B. |
| 269716 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARMD14, CO2 | |
| C2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title