missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C2b Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1936
This item is not returnable.
View return policy
Description
Complement Component C2b Monoclonal specifically detects Complement Component C2b in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Complement Component C2b | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P06681 | |
| C2 | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Component C2 Ala21-Leu752 Accession # P06681 | |
| 100 μg | |
| Primary | |
| Detects human Complement Component C2b in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) C1r, rhC1s, rhC3r, rhC5a, rhFactor B, or rhFactor I is observed. In Western blots, this antibody recognizes rhC2 and rhC2B. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 269716 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARMD14, CO2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 717 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction