missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C1qA, Mouse anti-Human, Clone: C1QA/2783, Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £405.00
Specifications
| Antigen | Complement Component C1qA |
|---|---|
| Clone | C1QA/2783 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, ELISA, Protein Array |
| Applications | Western Blot, ELISA, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18279947
|
Novus Biologicals
NBP2-79781-20UG |
20 μg |
£221.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18255227
|
Novus Biologicals
NBP2-79781-100UG |
100 μg |
£405.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Component C1qA Monoclonal specifically detects Complement Component C1qA in Human samples. It is validated for Western Blot, ELISA, Protein Array.Specifications
| Complement Component C1qA | |
| 0.2 mg/mL | |
| Western Blot, ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity | |
| 10 mM PBS with 0.05% BSA | |
| C1QA complement component 1, q subcomponent, A chain | |
| C1QA | |
| IgG2b κ | |
| Protein A or G purified |
| C1QA/2783 | |
| Western Blot 1-2 ug/ml, ELISA, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P02745, P02745 | |
| 712 | |
| Recombinant fragment (around aa 104-237) of human Complement Component C1qA protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title