missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C1qA, Mouse anti-Human, Clone: C1QA/2783, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79781-20UG
175.10 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Complement Component C1qA Monoclonal specifically detects Complement Component C1qA in Human samples. It is validated for Western Blot, ELISA, Protein Array.
Specifications
| Complement Component C1qA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA, Protein Array | |
| P02745, P02745 | |
| C1QA | |
| Recombinant fragment (around aa 104-237) of human Complement Component C1qA protein (exact sequence is proprietary) | |
| 20 μg | |
| Immunology, Innate Immunity | |
| 712 | |
| Store at 4°C. | |
| IgG2b κ |
| Western Blot, ELISA, Peptide Array | |
| C1QA/2783 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| C1QA complement component 1, q subcomponent, A chain | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction