missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Complement C4a Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£161.00 - £341.00
Specifications
| Antigen | Complement C4a |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17949652
|
Invitrogen™
PA5145474 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17929942
![]() |
Invitrogen™
PA5145473 |
200 μL |
£341.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement C4a is a protein that is part of the complement system, which is a group of proteins that work together to help the immune system fight off infections. C4a is produced when the complement system is activated, and it plays a role in inflammation and the recruitment of immune cells to the site of infection. It is also involved in the clearance of immune complexes and the regulation of the complement system.Specifications
| Complement C4a | |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| P08649 | |
| 24233, 625018 | |
| Recombinant Rat C4a protein, Asn678-Arg753 (Accession #P08649). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| C4A; C4AD; CO4A; Complement C4-A | |
| C4a | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| C4a |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title