missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Complement C4a Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145474
This item is not returnable.
View return policy
Description
Complement C4a is a protein that is part of the complement system, which is a group of proteins that work together to help the immune system fight off infections. C4a is produced when the complement system is activated, and it plays a role in inflammation and the recruitment of immune cells to the site of infection. It is also involved in the clearance of immune complexes and the regulation of the complement system.
Specifications
| Complement C4a | |
| Polyclonal | |
| Unconjugated | |
| C4a | |
| C4A; C4AD; CO4A; Complement C4-A | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 24233, 625018 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P08649 | |
| C4a | |
| Recombinant Rat C4a protein, Asn678-Arg753 (Accession #P08649). | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction