Learn More
R&D Systems™ CometSlide
Product Code. 18402050
Change view
Click to view available options
:
100 Slides
Product Code.
18402050 100 Slides
1 options
Product Code. 18402050 Supplier R&D Systems™ Supplier No. 425020003
Sample-ready pretreated glass slides used for the rapid analysis of DNA fragmentation associated with DNA damage
Convenient, simple and reliable sample preparation, processing, analysis and storage format for the CometAssay kits
Features a writing surface, specially treated glass surface and hydrophobic barrier for sample
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 Slides