missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 2mm Thick

Product Code. 11897583 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric) Well:
40 μL
50 μL
70 μL
80 μL
100 μL
120 μL
200 μL
1300 μL
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Well Unit Size
11897583 80 μL Each
11827593 1300 μL Each
11877593 70 μL Each
11827603 50 μL Each
11877603 40 μL Each
11827613 200 μL Each
11877613 120 μL Each
11827623 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11897583
  • ONLINE EXCLUSIVE PRICE
    £19.20
    £18.71 / Pack of 1
    Save £0.49
    2% Off
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

For use in preparatory electrophoresis and high-throughput screening.

  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • Recommended for maximising sample volume
  • 8-sample and 16-sample combs are Multichannel pipettor compatible

Specifications

Color Blue
For Use With (Application) Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations
For Use With (Equipment) OmniPAGE Mini Vertical Gel Electrophoresis Unit
Type Comb
No. of Wells 10
Quantity 1 Each
Thickness (Metric) 2 mm
Volume (Metric) Well 80 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.