Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 1.5mm Thick
Product Code. 11887583 Shop All Fisherbrand Products
Click to view available options
No. of Wells:
1 Prep, 1 Marker
12
16
20
5
88
9
Volume (Metric) Well:
30 μL
40 μL
50 μL
60 μL
70 μL
80 μL
140 μL
1000 μL
Unit Size:
Each
Product Code. 11887583
Brand: Fisherbrand™ VS10101.5
Only null left
Add to basket
Add to basket
For use in preparatory electrophoresis and high-throughput screening.
  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • Recommended for maximising sample volume
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
For Use With (Application) Mini SDS PAGE, Native PAGE, Gradient, Second Dimension and Nucleic Acid Separations
For Use With (Equipment) OmniPAGE Mini Vertical Gel Electrophoresis Unit
Type Comb
No. of Wells 12
Quantity 1 Each
Thickness (Metric) 1.5 mm
Volume (Metric) Well 60 μL