missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for HU13 Midi Horizontal Gel Electrophoresis Unit, 1mm thick

For use with HU13 Midi Horizontal Gel Electrophoresis Unit

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.01800kg

 View more versions of this product

Product Code. 10633812

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Color coded
  • Height adjustable
  • Offer complete control over loading volume
  • Well depth to maximum 112-sample throughput
Specifications

Specifications

HU13 Midi Horizontal Gel Electrophoresis Unit
28
13 μL
Comb
1 mm
3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers