missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 0.75mm Thick

For use in preparatory electrophoresis and high-throughput screening.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11873363

  • £21.00 / Each
Estimated Shipment: 31-07-2024
to see stock.
Add to basket

Description

Description

  • For tightly resolved bands
  • Recommended for high resolution gels and publication quality data
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications

Specifications

Black
Comb
1 Each
22 μL
MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit
10
0.75 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers