Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 0.75mm Thick
For use in preparatory electrophoresis and high-throughput screening.
£21.00 - £33.40
Specifications
For Use With (Equipment) | MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit |
---|---|
Type | Comb |
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
11803393
|
Fisherbrand™
|
2 Prep, 2 Marker | 169 μL |
£21.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11823383
|
Fisherbrand™
|
16 | 20 μL |
£21.95
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11873363
|
Fisherbrand™
|
10 | 22 μL |
£21.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11883373
|
Fisherbrand™
|
14 | 22 μL |
£21.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11843373
|
Fisherbrand™
|
12 | 30 μL |
£21.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11823363
|
Fisherbrand™
|
1 Prep, 1 Marker | 371 μL |
£21.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11833363
|
Fisherbrand™
|
10 | 34 μL |
£21.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11863403
|
Fisherbrand™
|
35 | 7 μL |
£21.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11863383
|
Fisherbrand™
|
18 | 11 μL |
£33.35
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
11813393
|
Fisherbrand™
|
20 | 16 μL |
£33.40
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
11883393
|
Fisherbrand™
|
28 | 8 μL |
£21.65
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11823403
|
Fisherbrand™
|
30 | 9 μL |
£21.00
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
11803413
|
Fisherbrand™
|
4 Prep, 2 Marker | 91 μL |
£21.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
Description
- For tightly resolved bands
- Recommended for high resolution gels and publication quality data
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit | |
Comb |