missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for Easigel H1-SET Horizontal Gel Electrophoresis Unit

For use with Easigel H1-SET Horizontal Gel Electrophoresis Unit

£39.30 - £69.80

Specifications

For Use With (Equipment) Easigel H1-SET Horizontal Gel Electrophoresis Unit
Type Comb
View More Specs

Products 7
Product Code Brand Width (Metric) Teeth Volume (Metric) Sample No. of Wells Thickness (Metric) Price Quantity & Availability  
Product Code Brand Width (Metric) Teeth Volume (Metric) Sample No. of Wells Thickness (Metric) Price Quantity & Availability  
11766874
View Documents Promotion Details
Fisherbrand™
H1C112
6 mm 27 μL 12 1 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10593944
View Documents Promotion Details
Fisherbrand™
H1-C1-16
4 mm 18 μL 16 1 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11760545
View Documents Promotion Details
Fisherbrand™
H1C120MC
2.8 mm 12 μL 20 1 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11776874
View Documents Promotion Details
Fisherbrand™
H1C1512
6 mm 40 μL 12 1.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11740545
View Documents Promotion Details
Fisherbrand™
H1C18
10 mm 45 μL 8 1 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11710555
View Documents Promotion Details
Fisherbrand™
H1C1520MC
2.8 mm 18 μL 20 1.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11790545
View Documents Promotion Details
Fisherbrand™
H1C158
10 mm 65 μL 8 1.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Color coded
  • Offer flexibility in sample-loading
  • Volume to maximum 40-sample throughput
Specifications

Specifications

Easigel H1-SET Horizontal Gel Electrophoresis Unit
Comb
Videos
SDS
Documents

Documents

Product Certifications