Learn More
Fisherbrand™ Comb for Wide Format Mini-Plus Horizontal Electrophoresis System
Product Code. 11573372 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
No. of Wells:
20
Unit Size:
Each
Product Code. No. of Wells Unit Size
11573372 20 Each
1 options
Product Code. 11573372
  • ONLINE EXCLUSIVE PRICE
    £37.90
    £32.56 / Pack of 1
    Save £5.34
    14% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Use with Wide Format Mini-Plus Horizontal Electrophoresis System.
  • Use with wide format Mini-Plus system
Specifications
For Use With (Equipment) Wide Format Mini-Plus Horizontal Gel Unit
Type Comb
No. of Wells 20
Thickness (English) 0.06 in.
Thickness (Metric) 1.5 mm
Volume (Metric) Sample 32 μL