missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Comb Block for multiSUB™ Midi 96 Gel System

Product Code. 12327977 Shop All Fisherbrand Products
Change view
Click to view available options
Thickness (Metric):
Comb block: 1 mm
Comb block: 1.5 mm
Includes:
1 Marker, Comb Block
2 Marker, Comb Block
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Includes unitSize
12327977 Comb block: 1.5 mm 2 Marker, Comb Block Each
12317977 Comb block: 1.5 mm 1 Marker, Comb Block Each
12307977 Comb block: 1 mm 1 Marker, Comb Block Each
12337977 Comb block: 1 mm 2 Marker, Comb Block Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12327977
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provides convenient loading template for single and multi-channel pipettes.

  • Fits virtually every application from preparatory electrophoresis to high-throughput screening
  • Height-adjustable without any requirement for specialist tools or comb holders
  • Prevents heat-induced warping

Specifications

For Use With (Equipment) multiSUB™ Midi96 Gel System
Type Comb Block
Format 96-well
Includes 2 Marker, Comb Block
Sample Size 8
Thickness (Metric) Comb block: 1.5 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.