missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COM1/p8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £309.00
Specifications
| Antigen | COM1/p8 |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
COM1/p8 Polyclonal specifically detects COM1/p8 in Human samples. It is validated for Western Blot.Specifications
| COM1/p8 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Candidate of metastasis 1, COM1P8, nuclear protein 1, nuclear protein, transcriptional regulator, 1, p8, Protein p8 | |
| NUPR1 | |
| IgG | |
| Affinity Purified |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis | |
| PBS with 0.02% Sodium Azide | |
| 26471 | |
| Reacts with the C terminus of the human p8 protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title