missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COM1/p8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-23627
This item is not returnable.
View return policy
Description
COM1/p8 Polyclonal specifically detects COM1/p8 in Human samples. It is validated for Western Blot.
Specifications
| COM1/p8 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| Candidate of metastasis 1, COM1P8, nuclear protein 1, nuclear protein, transcriptional regulator, 1, p8, Protein p8 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| NUPR1 | |
| Reacts with the C terminus of the human p8 protein. | |
| 0.1 mg | |
| Apoptosis | |
| 26471 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction