missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Columbia Agar with Sheep Blood PLUS

Product Code. 10463833
Change view
Click to view available options
Quantity:
10/Pk.
Format:
90 mm Monoplate
90mm Monoplate
Media Type:
Agar with 5% Sheep Blood
Agar with Sheep Blood
Manufacturing Origin:
Germany
Perth, UK
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Media Type Manufacturing Origin Unit Size
10463833 10/Pk. 90mm Monoplate Agar with 5% Sheep Blood Germany Pack of 10
12947128 10/Pk. 90 mm Monoplate Agar with Sheep Blood Perth, UK Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10463833 Supplier Thermo Scientific™ Supplier No. PB5039A
  • ONLINE EXCLUSIVE PRICE
    £12.95
    £12.70 / Pack of 10
    Save £0.25
    1% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and cultivate fastidious microorganisms with clearly visible haemolytic reactions (staphylococci and streptococci) with a medium containing sheep blood.

Isolate and cultivate fastidious microorganisms with clearly visible haemolytic reactions (Staphylococci and Streptococci) with a media formulation that contains sheep blood. Thermo Scientific™ Columbia Agar with Sheep Blood PLUS promotes haemolysis typical of Staphylococcus aureus, and it gives typical growth for Streptococcus pneumonia (dent morphology).

Grow fastidious organisms with rapid production of large colonies, clearly defined zones of haemolysis and good colonial differentiation for all-around performance. Traditionally, blood agar has been either casein hydrolysate or meat infusion media. The advantage of the first lies in the rapid production of large colonies, and the second in clearly defined zones of haemolysis and good colonial differentiation. This formulation combines the virtues of both for optimal performance.

Staphylococcus aureus
is a coagulase-positive, catalase positive, gram-positive cocci that occurs singly or in pairs, tetrads, short chains, and grape-like clusters. It is commonly carried on the skin and in the nose of healthy individuals but is also a major cause of infection in both healthcare and community settings worldwide. S. aureus can produce a variety of syndromes with clinical manifestations including skin and soft tissue infections, empyema, bloodstream infections, pneumonia, osteomyelitis, septic arthritis, endocarditis, sepsis, and meningitis.

Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta or gamma according to the appearance of zones around colonies growing on blood agar. β-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Traffic red, opaque
Content And Storage 2° to 12°C, in the dark
Format 90mm Monoplate
Manufacturing Origin Germany
Media Type Agar with 5% Sheep Blood
Product Type Agar Plate
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.