missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Columbia Agar with Sheep Blood PLUS / Brilliance™ MRSA 2 Agar

Codice prodotto. 15813237
Cambia vista
Click to view available options
Description:
Columbia Agar with Sheep Blood PLUS/BrillianceMRSA 2 Agar
Quantity:
10/Pk.
Manufacturing Origin:
Germany
Dimensione della confezione:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Description Quantity Manufacturing Origin unitSize
15813237 Columbia Agar with Sheep Blood PLUS/BrillianceMRSA 2 Agar 10/Pk. Germany Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15813237 Fornitore Thermo Scientific™ N. del fornitore PB5253E

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Grow fastidious organisms with clear haemolytic reactions (such as staphylococci) while screening for the presence of Methicillin-resistant Staphylococcus aureus (MRSA).

Isolate and cultivate fastidious microorganisms with clearly visible haemolytic reactions (staphylococci and streptococci), while simultaneously screening clinical samples for the presence of Methicillin-resistant Staphylococcus aureus (MRSA), using Thermo Scientific™ Columbia Agar with Sheep Blood PLUS / Brilliance™ MRSA 2 Agar, a single bi-plate that includes two separate media formulations. Identify MRSA and other organisms from one patient sample, plus verify samples are taken correctly if the MRSA result is negative.

The Columbia Agar with blood formulation on one side of the plate contains 5% sheep blood to promote growth of fastidious organisms with clearly visible haemolysis forms for staphylococci and streptococci, and it gives typical growth for S. pneumoniae (dent morphology).

Brilliance MRSA 2 Agar on the other side of the plate allows you to obtain faster results for the presence of MRSA with no need for reincubation of negatives. Get more reliable, more accurate results with this enhanced media formulation which contains two chromogens that differentiate MRSA and non-MRSA by distinctive blue colonies and produces larger MRSA colony growth compared to previous formulations.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifica

Color MRSA2: Light ivory, opaque. COL+SB: Traffic red, opaque
Content And Storage 2° to 12°C, in the dark
Description Columbia Agar with Sheep Blood PLUS/BrillianceMRSA 2 Agar
Format 90mm bi-plate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.