Learn More
Thermo Scientific™ Columbia Agar with Chocolated Horse Blood and Bacitracin
Description
Isolate Haemophilus species from clinical specimens with this ready-to-use medium.
Isolate Haemophilus spp. from clinical specimens with Thermo Scientific™ Oxoid™ Columbia Agar with Chocolated Horse Blood and Bacitracin. Chocolated horse blood is added to the general purpose Columbia Agar Base to supply the required nutrients for the growth of Haemophilus. Bacitracin is added as a selective agent to aid isolation of Haemophilus influenzae and overcome overgrowth of competing flora from the respiratory tract.
Use Columbia Agar with Chocolated Horse Blood and Bacitracin for the isolation of Haemophilus spp. from clinical specimens.
- Supports the growth of Haemophilus with chocolated blood providing the nutritional requirements.
- Improved isolation of Haemophilus influenzae due to bacitracin reducing competing respiratory flora.
- Ready-to-use convenience of prepared media.
It has been shown that isolation rates are not affected by the addition of bacitracin; however the quantity of H. influenzae growth increased due to reduced competing flora1.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Color | Brown |
| Content And Storage | 2°C to 10°C |
| Description | Columbia Agar with Chocolated Horse Blood and Bacitracin |
| Format | 90mm Monoplate |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar Plate |
| Fill Weight | 19.5g±1.0.g |
| Packaging Type | 10 x 90mm plates, film wrapped |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.