missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Columbia Agar with Blood / MacConkey Agar No. 3

Product Code. 10727553
Change view
Click to view available options
Description:
Columbia Agar with Blood / MacConkey Agar No. 3
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity unitSize
10727553 Columbia Agar with Blood / MacConkey Agar No. 3 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10727553 Supplier Thermo Scientific™ Supplier No. PB5207E
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Simultaneously grow fastidious organisms such as staphylococci and streptococci, detect and enumerate coliforms and isolate and detect Salmonella and Shigella spp.

Isolate and cultivate fastidious microorganisms with clearly visible haemolytic reactions (staphylococci and streptococci), detect and enumerate coliform organisms such as E. coli, while at the same time isolating and detecting Salmonella and Shigella spp., using Thermo Scientific™ Columbia Agar with Blood / MacConkey Agar No. 3, a single plate that includes two media formulations.

Read more results from a single patient sample and produce more diagnostic details of pathogenic organisms whilst reducing storage capacity issues in the incubator with a bi-plate that includes two widely used microbiological media.

Columbia Agar with Blood on one side of the plate contains 7% sheep blood to promote growth of fastidious organisms with clearly visible haemolysis forms for staphylococci and streptococci, and it gives typical growth for S.pneumoniae (dent morphology).

MacConkey Agar No. 3 on the other side of the plate is a selective medium for differentiating between coliforms and lactose and non-lactose fermenters while gram-positive cocci are completely inhibited.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color COL + SB: Carmine red, opaque. MAC3: Antique red, transparent
Content And Storage 2° to 12°C, in the dark
Description Columbia Agar with Blood / MacConkey Agar No. 3
Format 90mm biplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.