missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen XXI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | Collagen XXI |
|---|---|
| Concentration | 0.05mg/mL |
| Dilution | Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18499141
|
Novus Biologicals
NBP1-91015-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18347440
|
Novus Biologicals
NBP1-91015 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Collagen XXI Polyclonal specifically detects Collagen XXI in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Collagen XXI | |
| Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Cytoskeleton Markers, Extracellular Matrix | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| alpha 1 chain-like collagen, alpha 1 type XXI collagen, COL1AL, COLA1L, Collagen 21, collagen alpha-1(XXI) chain, collagen, type XXI, alpha 1, Collagen-21, dJ682J15.1, dJ708F5.1, DKFZp564B052, FLJ39125, FLJ44623, MGC26619 | |
| COL21A1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.05mg/mL | |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| Q96P44 | |
| 81578 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AKSSRFLTKIAVVLTDGKSQDDVKDAAQAARDSKITLFAIGVGSETEDAELRAIANKPSSTYVFYVEDYIAISKIREVMK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title