missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VI alpha 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£378.00
Specifications
| Antigen | Collagen VI alpha 2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
Collagen VI alpha 2 Polyclonal specifically detects Collagen VI alpha 2 in Human samples. It is validated for Western Blot.Specifications
| Collagen VI alpha 2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| collagen alpha-2(VI) chain, collagen VI, alpha-2 polypeptide, collagen, type VI, alpha 2,10PP3610, DKFZp586E1322, FLJ46862 | |
| COL6A2 | |
| IgG | |
| Protein A purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Q9BUM6 | |
| 1292 | |
| Synthetic peptides corresponding to COL6A2(collagen, type VI, alpha 2) The peptide sequence was selected from the C terminal of COL6A2. Peptide sequence ARRFVEQVARRLTLARRDDDPLNARVALLQFGGPGEQQVAFPLSHNLTAI. | |
| Primary | |
| 109 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title