missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VI alpha 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54614
This item is not returnable.
View return policy
Description
Collagen VI alpha 2 Polyclonal specifically detects Collagen VI alpha 2 in Human samples. It is validated for Western Blot.
Specifications
| Collagen VI alpha 2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| collagen alpha-2(VI) chain, collagen VI, alpha-2 polypeptide, collagen, type VI, alpha 2,10PP3610, DKFZp586E1322, FLJ46862 | |
| Rabbit | |
| 109 kDa | |
| 100 μL | |
| Primary | |
| Rat: 86%;. | |
| Human, Rat, Pig, Bovine, Canine, Equine | |
| Purified |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9BUM6 | |
| COL6A2 | |
| Synthetic peptides corresponding to COL6A2(collagen, type VI, alpha 2) The peptide sequence was selected from the C terminal of COL6A2. Peptide sequence ARRFVEQVARRLTLARRDDDPLNARVALLQFGGPGEQQVAFPLSHNLTAI. | |
| Protein A purified | |
| RUO | |
| 1292 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction