missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VI alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-59126
This item is not returnable.
View return policy
Description
Collagen VI alpha 1 Polyclonal specifically detects Collagen VI alpha 1 in Human samples. It is validated for Western Blot.
Specifications
| Collagen VI | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Collagen 6, collagen alpha-1(VI) chain, collagen VI, alpha-1 polypeptide, collagen, type VI, alpha 1, Collagen-6, human mRNA for collagen VI alpha-1 C-terminal globular domain10alpha 1 (VI) chain (61 AA), OPLL | |
| Rabbit | |
| 108 kDa | |
| 100 μL | |
| Cytoskeleton Markers, Extracellular Matrix | |
| 1291 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P12109 | |
| COL6A1 | |
| Synthetic peptides corresponding to COL6A1(collagen, type VI, alpha 1) The peptide sequence was selected from the middle region of COL6A1 (NP_001839). Peptide sequence ADITILLDGSASVGSHNFDTTKRFAKRLAERFLTAGRTDPAHDVRVAVVQ. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Human: 100%; Pig: 100%; Rabbit: 100%; Bovine: 92%; Equine: 92%; Mouse: 85%; Rat: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction